Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGA-2727 | 666842-36-0 | 98.8% | 536.62 g·mol⁻¹ | C26H40N4O8 | 100MG
SDP

Catalog No. 5000236805
Encompass_Preferred

KGA-2727 is a small-molecule, orally active, selective inhibitor of the sodium-glucose cotransporter SGLT1 used as a research compound to investigate glucose transport and metabolic effects. Reported Ki values are 97.4 nM for human SGLT1 and 43.5 nM for rat SGLT1. The compound is supplied as a solid (powder) for in vitro and in vivo studies; purity and pack sizes vary by supplier and batch.

  • Selective SGLT1 inhibitor; human Ki 97.4 nM, rat Ki 43.5 nM.
  • CAS number 666842-36-0.
  • Molecular formula C26H40N4O8; molecular weight 536.62 g·mol⁻¹.
  • Reported purity approximately 98.8% (supplier/batch dependent).
  • Typical form: solid (powder); multiple pack sizes available, including 100 mg.
  • Intended for research use to probe glucose transport and related metabolic pathways.

Catalog No. 50-002-36805 Supplier Medchemexpress LLC Supplier No. HY123797100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGA-2727 is a small-molecule, orally active, selective inhibitor of the sodium-glucose cotransporter SGLT1 used as a research compound to investigate glucose transport and metabolic effects. Reported Ki values are 97.4 nM for human SGLT1 and 43.5 nM for rat SGLT1. The compound is supplied as a solid (powder) for in vitro and in vivo studies; purity and pack sizes vary by supplier and batch.

  • Selective SGLT1 inhibitor; human Ki 97.4 nM, rat Ki 43.5 nM.
  • CAS number 666842-36-0.
  • Molecular formula C26H40N4O8; molecular weight 536.62 g·mol⁻¹.
  • Reported purity approximately 98.8% (supplier/batch dependent).
  • Typical form: solid (powder); multiple pack sizes available, including 100 mg.
  • Intended for research use to probe glucose transport and related metabolic pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.