Learn More
Medchemexpress LLC KGA-2727 | 666842-36-0 | MFCD32701917 | 98.8% | 536.62 | C26H40N4O8 | 25MG
SDP
Catalog No. 5000236829
Encompass_Preferred
KGA-2727 is a selective, high-affinity, orally active sodium-glucose cotransporter 1 (SGLT1) inhibitor used in preclinical research. It competitively inhibits human and rat SGLT1 (Ki 97.4 nM and 43.5 nM, respectively), shows substantial selectivity over SGLT2, and has demonstrated glucose-lowering efficacy in animal diabetes models.
  • Selective SGLT1 inhibition with competitive mechanism.
  • High affinity for human and rat SGLT1 (Ki ~97.4 nM and 43.5 nM).
  • Demonstrated glucose-lowering activity in preclinical diabetic models.
  • High reported purity suitable for research use.
  • Good solubility in DMSO and ethanol; moderate solubility in water with warming.
  • Solid form with defined storage recommendations for stability.
Catalog No. 50-002-36829 Supplier Medchemexpress LLC Supplier No. HY12379725MG
KGA-2727 is a selective, high-affinity, orally active sodium-glucose cotransporter 1 (SGLT1) inhibitor used in preclinical research. It competitively inhibits human and rat SGLT1 (Ki 97.4 nM and 43.5 nM, respectively), shows substantial selectivity over SGLT2, and has demonstrated glucose-lowering efficacy in animal diabetes models.
  • Selective SGLT1 inhibition with competitive mechanism.
  • High affinity for human and rat SGLT1 (Ki ~97.4 nM and 43.5 nM).
  • Demonstrated glucose-lowering activity in preclinical diabetic models.
  • High reported purity suitable for research use.
  • Good solubility in DMSO and ethanol; moderate solubility in water with warming.
  • Solid form with defined storage recommendations for stability.