Learn More
Medchemexpress LLC HY-P7047A 10ug Medchemexpress, KGF-1/FGF-7 (CHO-expressed), Human CAS: Purity:>98%
SDP
Catalog No. 501934293
Encompass_Preferred
Medchemexpress, HY-P7047A 10ug KGF-1/FGF-7 (CHO-expressed), Human CAS: Recombinant Human Keratinocyte Growth Factor 1/FGF-7 (CHO-expressed) is a polypeptide mitogen that belongs to the family of fibroblast growth factors, binds only to a splice variant of FGFR2 (FGFR2 IIIb) and is a highly specific paracrine growth factor for epithelial cells. Recombinant Human Keratinocyte Growth Factor 1/FGF-7 and its receptor are important for normal wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4293 Supplier Medchemexpress LLC Supplier No. HYP7047A10UG
Medchemexpress, HY-P7047A 10ug KGF-1/FGF-7 (CHO-expressed), Human CAS: Recombinant Human Keratinocyte Growth Factor 1/FGF-7 (CHO-expressed) is a polypeptide mitogen that belongs to the family of fibroblast growth factors, binds only to a splice variant of FGFR2 (FGFR2 IIIb) and is a highly specific paracrine growth factor for epithelial cells. Recombinant Human Keratinocyte Growth Factor 1/FGF-7 and its receptor are important for normal wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.