Learn More
Medchemexpress LLC KGF-2/FGF-10 Human 100ug
SDP
Catalog No. 5000184822
Encompass_Preferred
Recombinant human KGF-2/FGF-10 is a fibroblast growth factor used in research to stimulate epithelial cell proliferation, differentiation, and branching morphogenesis. Supplied lyophilized, this reagent is intended for in vitro receptor-binding studies and cell-based assays when reconstituted per the certificate of analysis.
  • Recombinant human protein, 169 amino acids.
  • Apparent molecular weight ~19-22 kDa by SDS-PAGE.
  • High purity (≥98%) as determined by reducing SDS-PAGE.
  • Expressed in Escherichia coli and supplied lyophilized.
  • Formulated from filtered PBS or Tris-HCl buffer for stability.
  • Biologically active in cell proliferation assays (ED50 ~9.6 ng/mL).
  • Stable when stored at -20°C; aliquot and freeze at -20°C/-80°C for long term.
  • Available in multiple microgram pack sizes for research use.
Catalog No. 50-001-84822 Supplier Medchemexpress LLC Supplier No. HYP7048100UG
Recombinant human KGF-2/FGF-10 is a fibroblast growth factor used in research to stimulate epithelial cell proliferation, differentiation, and branching morphogenesis. Supplied lyophilized, this reagent is intended for in vitro receptor-binding studies and cell-based assays when reconstituted per the certificate of analysis.
  • Recombinant human protein, 169 amino acids.
  • Apparent molecular weight ~19-22 kDa by SDS-PAGE.
  • High purity (≥98%) as determined by reducing SDS-PAGE.
  • Expressed in Escherichia coli and supplied lyophilized.
  • Formulated from filtered PBS or Tris-HCl buffer for stability.
  • Biologically active in cell proliferation assays (ED50 ~9.6 ng/mL).
  • Stable when stored at -20°C; aliquot and freeze at -20°C/-80°C for long term.
  • Available in multiple microgram pack sizes for research use.