Learn More
Medchemexpress LLC Kgf-2/fgf-10 protein, human (N-his) | >=95.0% | 10UG
SDP
Catalog No. 5000177464
Encompass_Preferred
Recombinant human KGF-2 (FGF-10) is an N-terminal His-tagged growth factor produced in Escherichia coli. It mediates epithelial-mesenchymal interactions, promotes epithelial proliferation and branching morphogenesis, and is used in research on tissue repair, cell signaling, and developmental biology. The product is provided as a purified recombinant protein suitable for laboratory applications.
  • N-His tagged recombinant human protein.
  • Expressed in Escherichia coli.
  • Approximately 21 kDa molecular weight.
  • Purity ≥95.0% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Useful for studies of epithelial proliferation, branching morphogenesis, and wound healing.
Catalog No. 50-001-77464 Supplier Medchemexpress LLC Supplier No. HYP70027910UG
Recombinant human KGF-2 (FGF-10) is an N-terminal His-tagged growth factor produced in Escherichia coli. It mediates epithelial-mesenchymal interactions, promotes epithelial proliferation and branching morphogenesis, and is used in research on tissue repair, cell signaling, and developmental biology. The product is provided as a purified recombinant protein suitable for laboratory applications.
  • N-His tagged recombinant human protein.
  • Expressed in Escherichia coli.
  • Approximately 21 kDa molecular weight.
  • Purity ≥95.0% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Useful for studies of epithelial proliferation, branching morphogenesis, and wound healing.