Learn More
Medchemexpress LLC HY-P7048 10ug Medchemexpress, KGF-2/FGF-10, Human CAS: Purity:>98%
SDP
Catalog No. 501934294
Encompass_Preferred
Medchemexpress, HY-P7048 10ug KGF-2/FGF-10, Human CAS: Recombinant Human Keratinocyte Growth Factor-2/FGF-10 is a heparin-binding protein that binds to FGF2 IIIb and FGFR1III-b receptors, promotes the growth, proliferation, and differentiation of epithelial cells and can enhance corneal wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4294 Supplier Medchemexpress LLC Supplier No. HYP704810UG
Medchemexpress, HY-P7048 10ug KGF-2/FGF-10, Human CAS: Recombinant Human Keratinocyte Growth Factor-2/FGF-10 is a heparin-binding protein that binds to FGF2 IIIb and FGFR1III-b receptors, promotes the growth, proliferation, and differentiation of epithelial cells and can enhance corneal wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.