Learn More
Medchemexpress LLC KGF-2/FGF-10 Human 20ug | 20ug
SDP
Catalog No. 5000187706
Encompass_Preferred
KGF-2/FGF-10 proteins coordinate embryonic development regulate cell proliferation and differentiation and are indispensable in branching morphogenesis This multifunctional protein may aid in wound healing KGF-2/FGF-10 Protein Human (Biotinylated) is the recombinant human-derived KGF-2/FGF-10 protein expressed by E coli with tag free
Catalog No. 50-001-87706 Supplier Medchemexpress LLC Supplier No. HYP7812320UG
KGF-2/FGF-10 proteins coordinate embryonic development regulate cell proliferation and differentiation and are indispensable in branching morphogenesis This multifunctional protein may aid in wound healing KGF-2/FGF-10 Protein Human (Biotinylated) is the recombinant human-derived KGF-2/FGF-10 protein expressed by E coli with tag free