Learn More
Medchemexpress LLC KGF-2/FGF-10 Human 2ug
SDP
Catalog No. 5000185004
Encompass_Preferred
Recombinant human Keratinocyte Growth Factor-2 (KGF-2/FGF-10) is a tag-free recombinant growth factor produced in Escherichia coli for research applications. It promotes epithelial cell proliferation, differentiation, and branching morphogenesis, and is used in studies of tissue repair, wound healing, and cell signaling.
  • Tag-free recombinant protein expressed in Escherichia coli.
  • Promotes epithelial cell proliferation and differentiation.
  • Supports branching morphogenesis in organotypic and developmental models.
  • Suitable for tissue repair, wound-healing, and cell signaling research.
  • Available in multiple pack sizes to match experimental scale.
  • Supplied for research use only.
Catalog No. 50-001-85004 Supplier Medchemexpress LLC Supplier No. HYP70482UG
Recombinant human Keratinocyte Growth Factor-2 (KGF-2/FGF-10) is a tag-free recombinant growth factor produced in Escherichia coli for research applications. It promotes epithelial cell proliferation, differentiation, and branching morphogenesis, and is used in studies of tissue repair, wound healing, and cell signaling.
  • Tag-free recombinant protein expressed in Escherichia coli.
  • Promotes epithelial cell proliferation and differentiation.
  • Supports branching morphogenesis in organotypic and developmental models.
  • Suitable for tissue repair, wound-healing, and cell signaling research.
  • Available in multiple pack sizes to match experimental scale.
  • Supplied for research use only.