Learn More
Medchemexpress LLC Recombinant human fibroblast growth factor 10 (FGF-10) | 2UG
SDP
Catalog No. 5000176288
Encompass_Preferred
Recombinant human fibroblast growth factor 10 (FGF-10), also known as KGF-2, is supplied as an N-terminal His-tagged, lyophilized protein for research use. It regulates epithelial-mesenchymal signaling, cell proliferation, and branching morphogenesis. The product is provided with low endotoxin and formulation/storage guidance in the manufacturer datasheet.
Catalog No. 50-001-76288 Supplier Medchemexpress LLC Supplier No. HYP7002792UG
Recombinant human fibroblast growth factor 10 (FGF-10), also known as KGF-2, is supplied as an N-terminal His-tagged, lyophilized protein for research use. It regulates epithelial-mesenchymal signaling, cell proliferation, and branching morphogenesis. The product is provided with low endotoxin and formulation/storage guidance in the manufacturer datasheet.