Learn More
Medchemexpress LLC KGF-2/FGF-10 Human 500ug
SDP
Catalog No. 5000184344
Encompass_Preferred
Recombinant human KGF-2/FGF-10 is a 169-amino-acid fibroblast growth factor provided as a purified, tag-free protein for cell biology research. It supports epithelial cell proliferation and branching morphogenesis and is commonly used in studies of development, wound healing, and growth factor signaling. Supplied in a 500 μg vial with reported high purity for in vitro applications.
  • 169 amino acids, matching native human sequence.
  • High purity (~98%) for consistent experimental results.
  • Tag-free format suitable for functional assays and receptor binding studies.
  • Expressed in E. coli and purified to remove contaminants.
  • Promotes epithelial cell proliferation and branching morphogenesis in vitro.
  • Supplied as 500 μg for multiple assay preparations.
Catalog No. 50-001-84344 Supplier Medchemexpress LLC Supplier No. HYP7048500UG
Recombinant human KGF-2/FGF-10 is a 169-amino-acid fibroblast growth factor provided as a purified, tag-free protein for cell biology research. It supports epithelial cell proliferation and branching morphogenesis and is commonly used in studies of development, wound healing, and growth factor signaling. Supplied in a 500 μg vial with reported high purity for in vitro applications.
  • 169 amino acids, matching native human sequence.
  • High purity (~98%) for consistent experimental results.
  • Tag-free format suitable for functional assays and receptor binding studies.
  • Expressed in E. coli and purified to remove contaminants.
  • Promotes epithelial cell proliferation and branching morphogenesis in vitro.
  • Supplied as 500 μg for multiple assay preparations.