Learn More
Medchemexpress LLC Fibroblast growth factor 10 (FGF-10), recombinant human | MFCD01867039 | ≥95.0% | 50UG
SDP
Catalog No. 5000176969
Encompass_Preferred
Recombinant human fibroblast growth factor 10 (FGF-10, KGF-2) is supplied as an N-terminal His-tagged, lyophilized protein produced in E. coli. It corresponds to residues L40-S208, has an approximate molecular weight of 21 kDa, and is intended for research into epithelial-mesenchymal signaling, cell proliferation, and branching morphogenesis.
  • High purity (≥95%) recombinant protein suitable for cell-based assays.
  • Low endotoxin level (<1 EU/μg) for reduced immunogenic interference.
  • Demonstrated biological activity: ED50 9.599-57 ng/mL; specific activity >5x10^4 units/mg.
  • Lyophilized powder for convenient long-term storage and transport.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for stability.
  • Stable stored at -20 °C; aliquot and keep at -20 °C or -80 °C for extended storage.
Catalog No. 50-001-76969 Supplier Medchemexpress LLC Supplier No. HYP70027950UG
Recombinant human fibroblast growth factor 10 (FGF-10, KGF-2) is supplied as an N-terminal His-tagged, lyophilized protein produced in E. coli. It corresponds to residues L40-S208, has an approximate molecular weight of 21 kDa, and is intended for research into epithelial-mesenchymal signaling, cell proliferation, and branching morphogenesis.
  • High purity (≥95%) recombinant protein suitable for cell-based assays.
  • Low endotoxin level (<1 EU/μg) for reduced immunogenic interference.
  • Demonstrated biological activity: ED50 9.599-57 ng/mL; specific activity >5x10^4 units/mg.
  • Lyophilized powder for convenient long-term storage and transport.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for stability.
  • Stable stored at -20 °C; aliquot and keep at -20 °C or -80 °C for extended storage.