Learn More
Medchemexpress LLC KGF-2/FGF-10 Human 5ug
SDP
Catalog No. 5000184673
Encompass_Preferred
Recombinant human KGF-2/FGF-10 (169 amino acids) is a biologically active growth factor involved in epithelial-mesenchymal interactions, branching morphogenesis, and regulation of cell proliferation and differentiation. This recombinant protein is provided for in vitro research applications including cell culture, signaling pathway studies, and developmental biology research.
  • Supports epithelial cell proliferation and differentiation.
  • Relevant for developmental biology and branching morphogenesis studies.
  • Suitable for cell culture and signaling assays.
  • Provided in microgram research quantities for experimental use.
  • Contains the human sequence of 169 amino acids.
Catalog No. 50-001-84673 Supplier Medchemexpress LLC Supplier No. HYP70485UG
Recombinant human KGF-2/FGF-10 (169 amino acids) is a biologically active growth factor involved in epithelial-mesenchymal interactions, branching morphogenesis, and regulation of cell proliferation and differentiation. This recombinant protein is provided for in vitro research applications including cell culture, signaling pathway studies, and developmental biology research.
  • Supports epithelial cell proliferation and differentiation.
  • Relevant for developmental biology and branching morphogenesis studies.
  • Suitable for cell culture and signaling assays.
  • Provided in microgram research quantities for experimental use.
  • Contains the human sequence of 169 amino acids.