Learn More
Medchemexpress LLC KGF-2/FGF-10 Mouse 100ug
SDP
Catalog No. 5000195915
Encompass_Preferred
KGF-2/FGF-10 Protein, Mouse is a recombinant growth factor that participates in paracrine signaling to regulate epithelial proliferation and branching morphogenesis in organs such as lung, skin, ear, and salivary glands. The product is supplied lyophilized with recommended formulations and storage instructions for reliable use in research applications.
  • Recombinant mouse FGF-10 spanning amino acids S62-T209.
  • Expressed in E. coli to support efficient production.
  • Lyophilized formulation for extended shelf life and convenient handling.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~17 kDa.
  • Stable when stored at -20°C; reconstituted aliquots can be frozen for long-term storage.
Catalog No. 50-001-95915 Supplier Medchemexpress LLC Supplier No. HYP7170100UG
KGF-2/FGF-10 Protein, Mouse is a recombinant growth factor that participates in paracrine signaling to regulate epithelial proliferation and branching morphogenesis in organs such as lung, skin, ear, and salivary glands. The product is supplied lyophilized with recommended formulations and storage instructions for reliable use in research applications.
  • Recombinant mouse FGF-10 spanning amino acids S62-T209.
  • Expressed in E. coli to support efficient production.
  • Lyophilized formulation for extended shelf life and convenient handling.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~17 kDa.
  • Stable when stored at -20°C; reconstituted aliquots can be frozen for long-term storage.