Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGF-2/FGF-10 Mouse 50ug
SDP

Catalog No. 5000183765
Encompass_Preferred

Recombinant mouse KGF-2/FGF-10 protein supplied lyophilized for research use. KGF-2/FGF-10 is a paracrine signaling molecule involved in branching morphogenesis in organs such as the lung, skin, ear, and salivary glands. The material is expressed in E. coli and supplied at research scales (for example, 50 μg). It is intended for in vitro applications such as cell proliferation assays.

  • Paracrine signaling molecule involved in branching morphogenesis of lung, skin, ear, and salivary glands.
  • Recombinant protein expressed in E. coli and provided as a lyophilized powder.
  • Predicted molecular mass approximately 17 kDa.
  • Recommended reconstitution concentration at or above 100 μg/mL in ddH2O.
  • Store frozen (-20°C) and aliquot for extended storage at -80°C.
  • Add carrier protein or trehalose for long-term stability.
  • For research use only; not for human or clinical use.

Catalog No. 50-001-83765 Supplier Medchemexpress LLC Supplier No. HYP717050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse KGF-2/FGF-10 protein supplied lyophilized for research use. KGF-2/FGF-10 is a paracrine signaling molecule involved in branching morphogenesis in organs such as the lung, skin, ear, and salivary glands. The material is expressed in E. coli and supplied at research scales (for example, 50 μg). It is intended for in vitro applications such as cell proliferation assays.

  • Paracrine signaling molecule involved in branching morphogenesis of lung, skin, ear, and salivary glands.
  • Recombinant protein expressed in E. coli and provided as a lyophilized powder.
  • Predicted molecular mass approximately 17 kDa.
  • Recommended reconstitution concentration at or above 100 μg/mL in ddH2O.
  • Store frozen (-20°C) and aliquot for extended storage at -80°C.
  • Add carrier protein or trehalose for long-term stability.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.