Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibroblast growth factor 10 (KGF-2), rat recombinant protein | >95.0% | 10UG
SDP

Catalog No. 5000175454
Encompass_Preferred

KGF-2/FGF-10 protein (rat) is a recombinant, tag-free fibroblast growth factor produced for research applications. It participates in epithelial-mesenchymal signaling that regulates embryonic development, cell proliferation, and branching morphogenesis, and is used in cell signaling and developmental biology studies. Refer to the supplier certificate of analysis for batch-specific purity and handling instructions.

  • Recombinant, rat-derived fibroblast growth factor 10.
  • Expressed in Escherichia coli and tag free.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Active in epithelial-mesenchymal signaling and branching morphogenesis assays.
  • Provided in research-scale quantities suitable for in vitro studies.

Catalog No. 50-001-75454 Supplier Medchemexpress LLC Supplier No. HYP70123210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGF-2/FGF-10 protein (rat) is a recombinant, tag-free fibroblast growth factor produced for research applications. It participates in epithelial-mesenchymal signaling that regulates embryonic development, cell proliferation, and branching morphogenesis, and is used in cell signaling and developmental biology studies. Refer to the supplier certificate of analysis for batch-specific purity and handling instructions.

  • Recombinant, rat-derived fibroblast growth factor 10.
  • Expressed in Escherichia coli and tag free.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Active in epithelial-mesenchymal signaling and branching morphogenesis assays.
  • Provided in research-scale quantities suitable for in vitro studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.