Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fibroblast growth factor 10 (FGF-10) | 62031-54-3 | >95.0% | 50UG
SDP

Catalog No. 5000175833
Encompass_Preferred

Recombinant rat fibroblast growth factor 10 (FGF-10, KGF-2) is a tag-free protein expressed in Escherichia coli representing the mature chain (Q37-S215). Supplied for research use, it supports studies of epithelial-mesenchymal signaling and branching morphogenesis. Typical analytical data indicate >95% purity by reducing SDS-PAGE and an observed molecular weight of approximately 23 kDa.

  • Tag-free mature chain corresponding to residues Q37-S215.
  • Expressed in Escherichia coli for consistent production.
  • High purity (>95%) suitable for cell culture and biochemical assays.
  • Observed molecular weight ~23 kDa, consistent with mature protein.
  • Intended for research use in developmental biology and signaling studies.

Catalog No. 50-001-75833 Supplier Medchemexpress LLC Supplier No. HYP70123250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat fibroblast growth factor 10 (FGF-10, KGF-2) is a tag-free protein expressed in Escherichia coli representing the mature chain (Q37-S215). Supplied for research use, it supports studies of epithelial-mesenchymal signaling and branching morphogenesis. Typical analytical data indicate >95% purity by reducing SDS-PAGE and an observed molecular weight of approximately 23 kDa.

  • Tag-free mature chain corresponding to residues Q37-S215.
  • Expressed in Escherichia coli for consistent production.
  • High purity (>95%) suitable for cell culture and biochemical assays.
  • Observed molecular weight ~23 kDa, consistent with mature protein.
  • Intended for research use in developmental biology and signaling studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.