Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGF/FGF-7 Human 500ug
SDP

Catalog No. 5000185521
Encompass_Preferred

Recombinant human KGF/FGF-7 is a tag-free polypeptide expressed in E. coli that stimulates proliferation and differentiation of epithelial cells. The material corresponds to residues C32-T194 (163 amino acids) of the human sequence, with a predicted molecular mass of ~18.9 kDa and >95% purity by reducing SDS-PAGE. Reconstitute at ≥100 μg/mL in ddH2O and store aliquots at -20°C or -80°C for long-term stability.

  • Promotes epithelial cell proliferation and differentiation.
  • Expressed in E. coli and provided tag free.
  • Contains 163 amino acids corresponding to residues C32-T194.
  • Predicted molecular mass ~18.9 kDa; purity >95% by reducing SDS-PAGE.
  • Stable when stored at -20°C; freeze aliquots at -80°C for extended storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Catalog No. 50-001-85521 Supplier Medchemexpress LLC Supplier No. HYP70597500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human KGF/FGF-7 is a tag-free polypeptide expressed in E. coli that stimulates proliferation and differentiation of epithelial cells. The material corresponds to residues C32-T194 (163 amino acids) of the human sequence, with a predicted molecular mass of ~18.9 kDa and >95% purity by reducing SDS-PAGE. Reconstitute at ≥100 μg/mL in ddH2O and store aliquots at -20°C or -80°C for long-term stability.

  • Promotes epithelial cell proliferation and differentiation.
  • Expressed in E. coli and provided tag free.
  • Contains 163 amino acids corresponding to residues C32-T194.
  • Predicted molecular mass ~18.9 kDa; purity >95% by reducing SDS-PAGE.
  • Stable when stored at -20°C; freeze aliquots at -80°C for extended storage.
  • Do not reconstitute below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.