Learn More
Medchemexpress LLC KGF/FGF-7 Human CH 2ug
SDP
Catalog No. 5000184672
Encompass_Preferred
Recombinant human keratinocyte growth factor (KGF/FGF-7), expressed in CHO cells and supplied as a lyophilized, tag-free preparation. This protein promotes proliferation and differentiation of epithelial cells and is intended for cell-based assays, receptor/ligand studies, and wound-healing research.
  • Expressed in CHO cells.
  • Tag free, full-length mature protein.
  • Glycosylated; apparent molecular weight ~16-27 kDa.
  • High purity (>95% by reducing SDS-PAGE).
  • Low endotoxin level (<0.2 EU/μg).
  • Potent biological activity (ED50 ~0.97 ng/mL).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage.
  • Reconstitute at ≥100 μg/mL in ddH2O and include carrier protein for prolonged stability.
Catalog No. 50-001-84672 Supplier Medchemexpress LLC Supplier No. HYP7047A2UG
Recombinant human keratinocyte growth factor (KGF/FGF-7), expressed in CHO cells and supplied as a lyophilized, tag-free preparation. This protein promotes proliferation and differentiation of epithelial cells and is intended for cell-based assays, receptor/ligand studies, and wound-healing research.
  • Expressed in CHO cells.
  • Tag free, full-length mature protein.
  • Glycosylated; apparent molecular weight ~16-27 kDa.
  • High purity (>95% by reducing SDS-PAGE).
  • Low endotoxin level (<0.2 EU/μg).
  • Potent biological activity (ED50 ~0.97 ng/mL).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage.
  • Reconstitute at ≥100 μg/mL in ddH2O and include carrier protein for prolonged stability.