Learn More
Medchemexpress LLC KGF/FGF-7 Human CH 50ug
SDP
Catalog No. 5000185353
Encompass_Preferred
KGF/FGF-7 Protein, Human (CHO) is a recombinant human keratinocyte growth factor produced in Chinese Hamster Ovary (CHO) cells. Supplied lyophilized, the protein is intended for research use in cell-based assays and biochemical studies where stimulation of epithelial cell proliferation and differentiation is required.
  • Mature sequence corresponding to human FGF-7 (C32-T194) for biological relevance.
  • Expressed in CHO cells to provide mammalian-type post-translational modifications.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Potent biological activity (ED50 ~0.97 ng/mL in a cell proliferation assay).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Recommend aliquoting and freezing for long-term storage.
Catalog No. 50-001-85353 Supplier Medchemexpress LLC Supplier No. HYP7047A50UG
KGF/FGF-7 Protein, Human (CHO) is a recombinant human keratinocyte growth factor produced in Chinese Hamster Ovary (CHO) cells. Supplied lyophilized, the protein is intended for research use in cell-based assays and biochemical studies where stimulation of epithelial cell proliferation and differentiation is required.
  • Mature sequence corresponding to human FGF-7 (C32-T194) for biological relevance.
  • Expressed in CHO cells to provide mammalian-type post-translational modifications.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Potent biological activity (ED50 ~0.97 ng/mL in a cell proliferation assay).
  • Lyophilized formulation with trehalose for enhanced stability.
  • Recommend aliquoting and freezing for long-term storage.