Learn More
Medchemexpress LLC KGF/FGF-7 Human HE 100ug
SDP
Catalog No. 5000184426
Encompass_Preferred
Recombinant human KGF/FGF-7 produced in HEK293 cells with a C-terminal His tag, supplied lyophilized for use in cell biology and tissue engineering. Intended for in vitro studies of epithelial cell proliferation, differentiation, and FGFR2 signaling, it is provided with storage and reconstitution guidance for research applications.
  • Produced in HEK293 mammalian cells for native-like glycosylation.
  • C-terminal 6xHis tag enables purification and detection.
  • High purity (≥90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose available for improved stability.
  • Recommended reconstitution at ≥100 μg/mL; add carrier protein for long-term storage.
  • Store frozen and use aliquots to avoid repeated freeze-thaw.
Catalog No. 50-001-84426 Supplier Medchemexpress LLC Supplier No. HYP70673100UG
Recombinant human KGF/FGF-7 produced in HEK293 cells with a C-terminal His tag, supplied lyophilized for use in cell biology and tissue engineering. Intended for in vitro studies of epithelial cell proliferation, differentiation, and FGFR2 signaling, it is provided with storage and reconstitution guidance for research applications.
  • Produced in HEK293 mammalian cells for native-like glycosylation.
  • C-terminal 6xHis tag enables purification and detection.
  • High purity (≥90%) as determined by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose available for improved stability.
  • Recommended reconstitution at ≥100 μg/mL; add carrier protein for long-term storage.
  • Store frozen and use aliquots to avoid repeated freeze-thaw.