Learn More
Medchemexpress LLC KGF/FGF-7 Human HE 10ug
SDP
Catalog No. 5000185069
Encompass_Preferred
Recombinant human keratinocyte growth factor (FGF-7) expressed in HEK293 cells with a His tag, provided in a small research pack for in vitro and cell-based studies. FGF-7 is a paracrine epithelial mitogen that promotes epithelial cell proliferation and differentiation; the recombinant format offers consistent material for signaling and bioassay applications.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Includes a C-terminal His tag to facilitate purification and detection.
  • Mature sequence corresponds to residues 32-194 (approximately 163 amino acids).
  • Approximate molecular weight suitable for signaling and bioactivity studies (≈19 kDa).
  • Suitable for cell-based assays and epithelial cell signaling studies.
  • Packaged in microgram-scale amounts for research use with manufacturer storage recommendations.
Catalog No. 50-001-85069 Supplier Medchemexpress LLC Supplier No. HYP7067310UG
Recombinant human keratinocyte growth factor (FGF-7) expressed in HEK293 cells with a His tag, provided in a small research pack for in vitro and cell-based studies. FGF-7 is a paracrine epithelial mitogen that promotes epithelial cell proliferation and differentiation; the recombinant format offers consistent material for signaling and bioassay applications.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Includes a C-terminal His tag to facilitate purification and detection.
  • Mature sequence corresponds to residues 32-194 (approximately 163 amino acids).
  • Approximate molecular weight suitable for signaling and bioactivity studies (≈19 kDa).
  • Suitable for cell-based assays and epithelial cell signaling studies.
  • Packaged in microgram-scale amounts for research use with manufacturer storage recommendations.