Learn More
Medchemexpress LLC KGF/FGF-7 Human HE 1mg
SDP
Catalog No. 5000184298
Encompass_Preferred
Recombinant human keratinocyte growth factor (FGF-7) produced in HEK293 cells and provided as a lyophilized His-tagged protein for in vitro research applications. It promotes epithelial cell proliferation and differentiation and is used in cell biology and signaling studies. Formulated in PBS (optionally with trehalose), the product shows an apparent molecular weight increase from glycosylation and is suitable for assays that require low endotoxin levels.
  • HEK293-expressed recombinant human protein.
  • His tag facilitates detection and purification.
  • Lyophilized formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Apparent molecular weight 20-28 kDa due to glycosylation.
  • Recommend reconstitution at ≥100 μg/mL and aliquot storage at -20°C or -80°C.
Catalog No. 50-001-84298 Supplier Medchemexpress LLC Supplier No. HYP706731MG
Recombinant human keratinocyte growth factor (FGF-7) produced in HEK293 cells and provided as a lyophilized His-tagged protein for in vitro research applications. It promotes epithelial cell proliferation and differentiation and is used in cell biology and signaling studies. Formulated in PBS (optionally with trehalose), the product shows an apparent molecular weight increase from glycosylation and is suitable for assays that require low endotoxin levels.
  • HEK293-expressed recombinant human protein.
  • His tag facilitates detection and purification.
  • Lyophilized formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Apparent molecular weight 20-28 kDa due to glycosylation.
  • Recommend reconstitution at ≥100 μg/mL and aliquot storage at -20°C or -80°C.