Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGF/FGF-7 Human HE 250ug
SDP

Catalog No. 5000184299
Encompass_Preferred

Recombinant human keratinocyte growth factor (KGF/FGF-7) produced in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. Intended for research use, it supports studies of epithelial cell proliferation, differentiation, and wound healing.

  • Recombinant human KGF expressed in HEK293 cells.
  • Supplied lyophilized for extended shelf life and convenient storage.
  • C-terminal 6xHis tag facilitates detection and purification.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Reconstitution guidance recommends concentrations of at least 100 μg/mL in water.
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage after reconstitution.

Catalog No. 50-001-84299 Supplier Medchemexpress LLC Supplier No. HYP70673250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human keratinocyte growth factor (KGF/FGF-7) produced in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. Intended for research use, it supports studies of epithelial cell proliferation, differentiation, and wound healing.

  • Recombinant human KGF expressed in HEK293 cells.
  • Supplied lyophilized for extended shelf life and convenient storage.
  • C-terminal 6xHis tag facilitates detection and purification.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Reconstitution guidance recommends concentrations of at least 100 μg/mL in water.
  • Store lyophilized at -20°C; aliquot and freeze for long-term storage after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.