Learn More
Medchemexpress LLC KGF/FGF-7 Human HE 2ug | 2ug
SDP
Catalog No. 5000184300
Encompass_Preferred
KGF/FGF-7 proteins coordinate embryonic development and regulate basic processes such as cell proliferation and differentiation Its critical role extends to normal branching morphogenesis KGF/FGF-7 Protein Human (HEK293 His) is the recombinant human-derived KGF/FGF-7 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-84300 Supplier Medchemexpress LLC Supplier No. HYP706732UG
KGF/FGF-7 proteins coordinate embryonic development and regulate basic processes such as cell proliferation and differentiation Its critical role extends to normal branching morphogenesis KGF/FGF-7 Protein Human (HEK293 His) is the recombinant human-derived KGF/FGF-7 protein expressed by HEK293 with C-6 His labeled tag