Learn More
Medchemexpress LLC KGF/FGF-7 Human HE 500ug
SDP
Catalog No. 5000185426
Encompass_Preferred
Recombinant human keratinocyte growth factor (KGF/FGF-7) expressed in HEK293 cells with a C-terminal His tag. The protein promotes epithelial cell proliferation and migration and is used in developmental biology, wound healing, epithelial differentiation, and organoid culture research.
  • Stimulates epithelial cell proliferation and migration.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag facilitates detection and purification.
  • Low endotoxin level suitable for sensitive bioassays.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.
  • Consistent construct corresponding to the mature protein for reproducible results.
Catalog No. 50-001-85426 Supplier Medchemexpress LLC Supplier No. HYP70673500UG
Recombinant human keratinocyte growth factor (KGF/FGF-7) expressed in HEK293 cells with a C-terminal His tag. The protein promotes epithelial cell proliferation and migration and is used in developmental biology, wound healing, epithelial differentiation, and organoid culture research.
  • Stimulates epithelial cell proliferation and migration.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • C-terminal His tag facilitates detection and purification.
  • Low endotoxin level suitable for sensitive bioassays.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.
  • Consistent construct corresponding to the mature protein for reproducible results.