Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGF/FGF-7 Human HE 50ug
SDP

Catalog No. 5000185076
Encompass_Preferred

Recombinant human keratinocyte growth factor (KGF/FGF-7), expressed in HEK293 cells with a C-terminal His tag, is supplied as a lyophilized 50 μg preparation for in vitro research. It promotes epithelial cell proliferation and differentiation and exhibits measurable bioactivity in multiple cell-based assays.

  • Expressed in mammalian cells to support proper folding and glycosylation.
  • Supplied as a lyophilized powder for improved stability and handling.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Demonstrated activity in multiple proliferation assays (ED50 in the ng/mL range).
  • Reconstitution at concentrations ≥100 μg/mL in ddH2O is recommended.
  • Store at -20°C; stable short-term at 4°C after reconstitution, aliquot for long-term storage.

Catalog No. 50-001-85076 Supplier Medchemexpress LLC Supplier No. HYP7067350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human keratinocyte growth factor (KGF/FGF-7), expressed in HEK293 cells with a C-terminal His tag, is supplied as a lyophilized 50 μg preparation for in vitro research. It promotes epithelial cell proliferation and differentiation and exhibits measurable bioactivity in multiple cell-based assays.

  • Expressed in mammalian cells to support proper folding and glycosylation.
  • Supplied as a lyophilized powder for improved stability and handling.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Demonstrated activity in multiple proliferation assays (ED50 in the ng/mL range).
  • Reconstitution at concentrations ≥100 μg/mL in ddH2O is recommended.
  • Store at -20°C; stable short-term at 4°C after reconstitution, aliquot for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.