Learn More
Medchemexpress LLC KGF/FGF-7 Human N- 100ug
SDP
Catalog No. 5000179172
Encompass_Preferred
KGF/FGF-7 Protein Human (His) is a polypeptide mitogen that belongs to the family of fibroblast growth factors binds only to a splice variant of FGFR2 (FGFR2 IIIb) and is a highly specific paracrine growth factor for epithelial cells KGF/FGF-7 Protein Human (His) and its receptor are important for normal wound healing
Catalog No. 50-001-79172 Supplier Medchemexpress LLC Supplier No. HYP7382100UG
KGF/FGF-7 Protein Human (His) is a polypeptide mitogen that belongs to the family of fibroblast growth factors binds only to a splice variant of FGFR2 (FGFR2 IIIb) and is a highly specific paracrine growth factor for epithelial cells KGF/FGF-7 Protein Human (His) and its receptor are important for normal wound healing