Learn More
Genscript Corporation Antibody Protein Proteins Proteins 5 UG 1/EA
Supplier:  Genscript Corporation Z031745
Encompass
Keratinocyte Growth Factor (KGF) is a highly specific epithelial mitogen produced by fibroblasts and mesenchymal stem cells. KGF belongs to the heparin binding Fibroblast Growth Factor (FGF) family and is known as FGF-7. However in contrast to the FGF-1 which binds to all known FGF receptors with high affinity KGF only binds to a splice variant of an FGF receptor FGFR2-IIIb. FGFR2-IIIb is produced by most of the epithelial cells indicating that KGF plays roles as a paracrine mediator. KGF induces the differen-tiation and proliferation of various epithelial cells including keratinocytes in the epidermis hair follicles and sebaceous glands and is responsible for the wound repairs of various tissues including lung bladder and kidney.
Catalog No. 50-252-9745