Learn More
Medchemexpress LLC KGF/FGF-7 Mouse 100ug
SDP
Catalog No. 5000195824
Encompass_Preferred
Recombinant mouse fibroblast growth factor 7 (FGF-7, keratinocyte growth factor) supplied as a lyophilized, tag-free protein expressed in E. coli. It is a potent epithelial mitogen used to stimulate cell proliferation and study signalling pathways in skin, intestinal, hepatic, pulmonary, and breast models. Supplied in microgram-scale pack sizes for research applications.
Catalog No. 50-001-95824 Supplier Medchemexpress LLC Supplier No. HYP7176100UG
Recombinant mouse fibroblast growth factor 7 (FGF-7, keratinocyte growth factor) supplied as a lyophilized, tag-free protein expressed in E. coli. It is a potent epithelial mitogen used to stimulate cell proliferation and study signalling pathways in skin, intestinal, hepatic, pulmonary, and breast models. Supplied in microgram-scale pack sizes for research applications.