Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGF/FGF-7 Mouse 2ug
SDP

Catalog No. 5000195534
Encompass_Preferred

KGF/FGF-7 Protein, Mouse is a recombinant, tag-free mouse fibroblast growth factor supplied lyophilized for research use. Produced in E. coli and purified to >95%, it promotes epithelial cell proliferation in tissues such as skin, intestine, breast, liver, and lung, and is intended for in vitro research applications.

  • Recombinant, tag-free mouse FGF-7 produced in E. coli.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Supplied lyophilized from PBS (pH 7.4) for convenient storage and handling.
  • Stable at -20°C for long-term storage; after reconstitution stable at 4°C for 1 week.
  • Available in small research sizes, including 2 μg, for scarce-sample experiments.
  • Promotes epithelial cell proliferation in multiple organ-derived cell types.
  • Sequence accession provided for identity confirmation (P36363, C32-T194).

Catalog No. 50-001-95534 Supplier Medchemexpress LLC Supplier No. HYP71762UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGF/FGF-7 Protein, Mouse is a recombinant, tag-free mouse fibroblast growth factor supplied lyophilized for research use. Produced in E. coli and purified to >95%, it promotes epithelial cell proliferation in tissues such as skin, intestine, breast, liver, and lung, and is intended for in vitro research applications.

  • Recombinant, tag-free mouse FGF-7 produced in E. coli.
  • Purified to greater than 95% as determined by reducing SDS-PAGE.
  • Supplied lyophilized from PBS (pH 7.4) for convenient storage and handling.
  • Stable at -20°C for long-term storage; after reconstitution stable at 4°C for 1 week.
  • Available in small research sizes, including 2 μg, for scarce-sample experiments.
  • Promotes epithelial cell proliferation in multiple organ-derived cell types.
  • Sequence accession provided for identity confirmation (P36363, C32-T194).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.