Learn More
Lonza Walkersville KBM Keratinocyte Basal Medium
Supplier:  Lonza Walkersville CC3101
Encompass
KBM is a basal medium sold separately for the proliferation of keratinocytes in a serum-free environment. This can be used with CC-4131 (keratinocyte growth medium SingleQuots Supplements and Growth Factors). Alternatively both components can be purchased in conjunction as KGM Keratinocyte Growth Medium BulletKit. Content:1 x Bottle KBM Basal Medium, 500 mL
Catalog No. NC9791321