Learn More
Lonza Walkersville KGM Keratinocyte Growth Medium BulletKit
Catalog No. NC9486029
Encompass
KGM Keratinocyte Growth Medium has been optimized for the proliferation of keratinocytes in a serum-free environment.Did you know our customers have used KGM Media also to culture immortalized corneal epithelial cells? Check out Jolly, et. al. publication that provides some guidelines to do this.Content:1 x KBM (CC-3101) Basal Medium, 500 mL1 x KGM (CC-4131) Singlequot Supplement Pack containing:1 x Orange Cap Vial with BPE, 2.00 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL
Catalog No. NC9486029 Supplier Lonza Walkersville Supplier No. CC3111
KGM Keratinocyte Growth Medium has been optimized for the proliferation of keratinocytes in a serum-free environment.Did you know our customers have used KGM Media also to culture immortalized corneal epithelial cells? Check out Jolly, et. al. publication that provides some guidelines to do this.Content:1 x KBM (CC-3101) Basal Medium, 500 mL1 x KGM (CC-4131) Singlequot Supplement Pack containing:1 x Orange Cap Vial with BPE, 2.00 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL