Learn More
Lonza Walkersville KGM Gold Keratinocyte Growth Medium BulletKit
Catalog No. NC1075051
Encompass
A new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.KGM-Gold Keratinocyte Growth Medium is designed to support both clonal growth and high density keratinocyte proliferation. The medium supports superior growth and morphology of keratinocytes isolated in KGM-2, KGM and Competitor A's Medium (data shown in the data tab).Content:1 x KBM Gold Basal Medium (00192151), 500 mL1 x KGMGold SingleQuots Supplement pack (00192152) containing: 1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Epinephrine, 0.25 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Orange Cap Vial with BPE, 2.00 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL
Catalog No. NC1075051 Supplier Lonza Walkersville Supplier No. 00192060
A new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.KGM-Gold Keratinocyte Growth Medium is designed to support both clonal growth and high density keratinocyte proliferation. The medium supports superior growth and morphology of keratinocytes isolated in KGM-2, KGM and Competitor A's Medium (data shown in the data tab).Content:1 x KBM Gold Basal Medium (00192151), 500 mL1 x KGMGold SingleQuots Supplement pack (00192152) containing: 1 x Natural Cap Vial with Hydrocortisone, 0.50 mL1 x Natural Cap Vial with Transferrin, 0.50 mL1 x Amber Vial with Epinephrine, 0.25 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Orange Cap Vial with BPE, 2.00 mL1 x Green Cap Vial with hEGF, 0.50 mL1 x Lilac Cap Vial with Insulin, 0.50 mL