Learn More
Lonza Walkersville KGM Gold BulletKit w/o Ca
Catalog No. 5000496471
Encompass
A robust and versatile serum-free medium formulation for high and low density NHEK-Normal human epidermal keratinocyte cultures KGM-Gold Calcium-Free Keratinocyte Growth Medium is designed to support both clonal growth and high density keratinocyte proliferation The medium supports superior growth and morphology of keratinocytes Furthermore quality control at Lonza requires that keratinocytes pass strict morphological criteria specifying the minimal presence of enlarged and vacuolated cells enabling Lonza keratinocytes to be used by the researcher for 15 population doublings
Catalog No. 50-004-96471 Supplier Lonza Walkersville Supplier No. 00195769
A robust and versatile serum-free medium formulation for high and low density NHEK-Normal human epidermal keratinocyte cultures KGM-Gold Calcium-Free Keratinocyte Growth Medium is designed to support both clonal growth and high density keratinocyte proliferation The medium supports superior growth and morphology of keratinocytes Furthermore quality control at Lonza requires that keratinocytes pass strict morphological criteria specifying the minimal presence of enlarged and vacuolated cells enabling Lonza keratinocytes to be used by the researcher for 15 population doublings