Learn More
Lonza Walkersville KGM Keratinocyte Growth Medium SingleQuots Supplements and Growth Factors
Catalog No. NC9566990
Encompass
KGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the KGM Keratinocyte Cell Growth Medium BulletKit (Catalog Number CC-3111) which consists of basal medium and supplements packaged together.
Catalog No. NC9566990 Supplier Lonza Walkersville Supplier No. CC4131
KGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the KGM Keratinocyte Cell Growth Medium BulletKit (Catalog Number CC-3111) which consists of basal medium and supplements packaged together.