Learn More
Biomatik Corporation Kgp Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11147
Encompass_Preferred
Kgp Polyclonal Antibody is a Porphyromonas gingivalis Polyclonal Antibody to kgp. The Kgp Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Porphyromonas gingivalis. Immunogen: Recombinant Porphyromonas Gingivalis Lys-Gingipain Protein (229-594Aa). Applications: ELISA, WB. Target Synonyms: kgp. Accession Number: B2RLK2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7624