Learn More
Medchemexpress LLC Kgp-IN-1 (hydrochloride) | 2097865-47-7 | 97.2% | 468.87 | C19H25ClF4N4O3 | 5 MG
SDP
Catalog No. 5000262834
Encompass_Preferred
Kgp-IN-1 hydrochloride is the hydrochloride salt of Kgp-IN-1, an arginine-specific gingipain (Rgp) inhibitor described in patent WO2017201322A1 (compound 13-R). It is supplied as a white to off-white solid for biochemical and pharmacological research, with reported purity of 97.18% and molecular weight 468.87 g·mol⁻¹.
  • Arginine-specific gingipain inhibitor useful for Rgp-targeted studies.
  • High purity suitable for in vitro research.
  • Available as solid and as pre-made DMSO solution formats.
  • White to off-white solid for reliable handling.
  • Structural and spectral identifiers provided for cheminformatics use.
Catalog No. 50-002-62834 Supplier Medchemexpress LLC Supplier No. HY128523A5MG
Kgp-IN-1 hydrochloride is the hydrochloride salt of Kgp-IN-1, an arginine-specific gingipain (Rgp) inhibitor described in patent WO2017201322A1 (compound 13-R). It is supplied as a white to off-white solid for biochemical and pharmacological research, with reported purity of 97.18% and molecular weight 468.87 g·mol⁻¹.
  • Arginine-specific gingipain inhibitor useful for Rgp-targeted studies.
  • High purity suitable for in vitro research.
  • Available as solid and as pre-made DMSO solution formats.
  • White to off-white solid for reliable handling.
  • Structural and spectral identifiers provided for cheminformatics use.