Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KGP94 (cathepsin L inhibitor) | 1131456-28-4 | 99.8% | 350.23 | C14H12BrN3OS | 5 MG
SDP

Catalog No. 5000228422
Encompass_Preferred

KGP94 is a selective small-molecule inhibitor of Cathepsin L for research use. It is provided in high-purity solid and solution formats for biochemical and cellular studies of cathepsin-mediated processes.

  • High purity (99.77%).
  • Molecular formula C14H12BrN3OS.
  • Molecular weight 350.23.
  • Supplied as a small solid dose suitable for in vitro studies.
  • Soluble in DMSO for solution preparation.

Catalog No. 50-002-28422 Supplier Medchemexpress LLC Supplier No. HY1187625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KGP94 is a selective small-molecule inhibitor of Cathepsin L for research use. It is provided in high-purity solid and solution formats for biochemical and cellular studies of cathepsin-mediated processes.

  • High purity (99.77%).
  • Molecular formula C14H12BrN3OS.
  • Molecular weight 350.23.
  • Supplied as a small solid dose suitable for in vitro studies.
  • Soluble in DMSO for solution preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.