Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KH-CB19 | 1354037-26-5 | 99.3% | 1 ML
SDP

Catalog No. 5000342888
Encompass_Preferred

KH-CB19 is a potent inhibitor of cdc2-like kinase (CLK), specifically targeting CLK1 with an IC50 of 19.7 nM and CLK3 with an IC50 of 530 nM. It also exhibits antiviral activity, inhibiting influenza virus replication with an IC50 of 13.6 μM. This compound is valuable as a research tool to study the regulation of alternative splicing.

  • Potent CLK inhibitor
  • Exhibits antiviral activity
  • Purity of 99.3%
  • Appears as a white to light yellow solid
  • Soluble in DMSO (≥ 50 mg/mL)
  • Molecular weight is 338.19
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-003-42888 Supplier Medchemexpress LLC Supplier No. HY1282810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KH-CB19 is a potent inhibitor of cdc2-like kinase (CLK), specifically targeting CLK1 with an IC50 of 19.7 nM and CLK3 with an IC50 of 530 nM. It also exhibits antiviral activity, inhibiting influenza virus replication with an IC50 of 13.6 μM. This compound is valuable as a research tool to study the regulation of alternative splicing.

  • Potent CLK inhibitor
  • Exhibits antiviral activity
  • Purity of 99.3%
  • Appears as a white to light yellow solid
  • Soluble in DMSO (≥ 50 mg/mL)
  • Molecular weight is 338.19
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.