Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KH-CB20 | 1354448-60-4 | 99.8% | 10 MG
SDP

Catalog No. 5000342885
Encompass_Preferred

KH-CB20, an E/Z mixture, is a potent and selective inhibitor of CLK1 (IC50 = 16.5 nM) and the closely related isoform CLK4. It also inhibits DYRK1A (IC50 = 57.8 nM) and CLK3 (IC50 = 488 nM). It is intended for research use only and not sold to patients.

  • Potent and selective CLK1 inhibitor
  • Inhibits CLK4, DYRK1A, and CLK3
  • Molecular weight: 338.19
  • Chemical formula: C15H13Cl2N3O2
  • Appearance: Solid, off-white to light yellow
  • For research use only

Catalog No. 50-003-42885 Supplier Medchemexpress LLC Supplier No. HY12828A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KH-CB20, an E/Z mixture, is a potent and selective inhibitor of CLK1 (IC50 = 16.5 nM) and the closely related isoform CLK4. It also inhibits DYRK1A (IC50 = 57.8 nM) and CLK3 (IC50 = 488 nM). It is intended for research use only and not sold to patients.

  • Potent and selective CLK1 inhibitor
  • Inhibits CLK4, DYRK1A, and CLK3
  • Molecular weight: 338.19
  • Chemical formula: C15H13Cl2N3O2
  • Appearance: Solid, off-white to light yellow
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.