Learn More
Medchemexpress LLC KH-CB20 | 1354448-60-4 | MFCD00407230 | 99.8% | 338.19 g/mol | C15H13Cl2N3O2 | 5 MG
SDP
Catalog No. 5000264645
Encompass_Preferred
KH-CB20 is a research-grade small-molecule inhibitor of CDC-like kinases (CLK1 and CLK4). Supplied as an E/Z isomer mixture, it exhibits potent biochemical activity and is provided as a high-purity solid for biochemical and cell-based assays related to splicing and cell-cycle regulation. Refer to the supplier datasheet for experimental and safety information.
  • Potent inhibition of CLK1 and CLK4 (CLK1 IC50 16.5 nM).
  • Also active against DYRK1A and CLK3.
  • High-purity solid suitable for analytical and preparative work.
  • Available in milligram quantities and as a DMSO solution.
  • Intended for research use only; follow safety and handling guidance.
Catalog No. 50-002-64645 Supplier Medchemexpress LLC Supplier No. HY12828A5MG
KH-CB20 is a research-grade small-molecule inhibitor of CDC-like kinases (CLK1 and CLK4). Supplied as an E/Z isomer mixture, it exhibits potent biochemical activity and is provided as a high-purity solid for biochemical and cell-based assays related to splicing and cell-cycle regulation. Refer to the supplier datasheet for experimental and safety information.
  • Potent inhibition of CLK1 and CLK4 (CLK1 IC50 16.5 nM).
  • Also active against DYRK1A and CLK3.
  • High-purity solid suitable for analytical and preparative work.
  • Available in milligram quantities and as a DMSO solution.
  • Intended for research use only; follow safety and handling guidance.