Learn More
Medchemexpress LLC HY-103194 10mg Medchemexpress, KH7 CAS:330676-02-3 Purity:>98%
SDP
Catalog No. 501968055
Encompass_Preferred
Medchemexpress, HY-103194 10mg KH7 CAS:330676-02-3 KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor, with IC50s of 3-10 μM toward both recombinant purified human sACt protein and heterologously expressed sACt in cellular assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8055 Supplier Medchemexpress LLC Supplier No. HY10319410MG
Medchemexpress, HY-103194 10mg KH7 CAS:330676-02-3 KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor, with IC50s of 3-10 μM toward both recombinant purified human sACt protein and heterologously expressed sACt in cellular assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.