Learn More
Aviva Systems Biology Corp KHDRBS2 Antibody - middle region (ARP53110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KHDRBS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KHDRBS2. Alternate names: FLJ38664, MGC26664, SLM-1, SLM1, bA535F17.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60924