Learn More
Aviva Systems Biology Corp KHDRBS3 Antibody - N-terminal region (ARP40664_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40664_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KHDRBS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KHDRBS3. Alternate names: Etle, SALP, SLM-2, SLM2, T-STAR, TSTAR, etoileApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53222