Learn More
Medchemexpress LLC Khellin | 82-02-0 | 99.7% | 5 MG
SDP
Catalog No. 5000411879
Encompass_Preferred
Khellin is a furochromone isolated from *Ammi visnuga L.* It acts as an EGFR inhibitor with an IC50 of 0.15 μM and exhibits anti-proliferative activity *in vitro*. Additionally, Khellin possesses antispasmodic and coronary vasodilator effects.
  • Acts as an EGFR inhibitor
  • Exhibits anti-proliferative activity in vitro
  • Possesses antispasmodic effects
  • Shows coronary vasodilator effects
Catalog No. 50-004-11879 Supplier Medchemexpress LLC Supplier No. HYB13945MG
Khellin is a furochromone isolated from *Ammi visnuga L.* It acts as an EGFR inhibitor with an IC50 of 0.15 μM and exhibits anti-proliferative activity *in vitro*. Additionally, Khellin possesses antispasmodic and coronary vasodilator effects.
  • Acts as an EGFR inhibitor
  • Exhibits anti-proliferative activity in vitro
  • Possesses antispasmodic effects
  • Shows coronary vasodilator effects