Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Khellin, MP Biomedicals

Catalog No. ICN202092 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202092 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202092 Supplier MP Biomedicals, Inc Supplier No. 0520209210
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.245
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
Synonym khellin, visammin, amicardine, methafrone, coronin, amikellin, ammipuran, ammivisnagen, benecardin, corafurone
PubChem CID 3828
ChEBI CHEBI:6133
IUPAC Name 4,9-dimethoxy-7-methylfuro[3,2-g]chromen-5-one
SMILES CC1=CC(=O)C2=C(C3=C(C(=C2O1)OC)OC=C3)OC

Specifications

Quantity 10 g
Formula Weight 260.24
Chemical Name or Material Khellin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.