Learn More
Medchemexpress LLC KHK-IN-1 hydrochloride | 1303470-48-5 | 98.0% | 50 MG
SDP
Catalog No. 5000342061
Encompass_Preferred
KHK-IN-1 hydrochloride is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor. It effectively inhibits the production of F1P in HepG2 cell lysates, demonstrating an IC50 value of 400 nM. This compound holds potential for research related to diabetes and obesity, offering a valuable tool for scientific investigation.
  • Demonstrates selectivity as a ketohexokinase inhibitor.
  • Is cell membrane permeable, allowing intracellular action.
  • Inhibits F1P production in HepG2 cell lysates.
  • Possesses potential for studying diabetes and obesity.
  • Stable in human and rat liver microsome preparations.
  • Does not significantly inhibit cytochrome P450s.
Catalog No. 50-003-42061 Supplier Medchemexpress LLC Supplier No. HY12841A50MG
KHK-IN-1 hydrochloride is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor. It effectively inhibits the production of F1P in HepG2 cell lysates, demonstrating an IC50 value of 400 nM. This compound holds potential for research related to diabetes and obesity, offering a valuable tool for scientific investigation.
  • Demonstrates selectivity as a ketohexokinase inhibitor.
  • Is cell membrane permeable, allowing intracellular action.
  • Inhibits F1P production in HepG2 cell lysates.
  • Possesses potential for studying diabetes and obesity.
  • Stable in human and rat liver microsome preparations.
  • Does not significantly inhibit cytochrome P450s.