Learn More
Medchemexpress LLC KHK-IN-1 hydrochloride | 1303470-48-5 | 98.47% | 10 MG
SDP
Catalog No. 5000341130
Encompass_Preferred
KHK-IN-1 hydrochloride is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor with an IC50 of 12 nM. It inhibits the production of F1P in HepG2 cell lysates with an IC50 of 400 nM and has potential for the study of diabetes and obesity.
  • Selective and cell membrane permeable KHK inhibitor
  • Inhibits F1P production in HepG2 cell lysates
  • Potential for diabetes and obesity research
  • Stable in human and rat liver microsome preparations
  • Does not significantly inhibit cytochrome P450s
Catalog No. 50-003-41130 Supplier Medchemexpress LLC Supplier No. HY12841A10MG
KHK-IN-1 hydrochloride is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor with an IC50 of 12 nM. It inhibits the production of F1P in HepG2 cell lysates with an IC50 of 400 nM and has potential for the study of diabetes and obesity.
  • Selective and cell membrane permeable KHK inhibitor
  • Inhibits F1P production in HepG2 cell lysates
  • Potential for diabetes and obesity research
  • Stable in human and rat liver microsome preparations
  • Does not significantly inhibit cytochrome P450s