Learn More
Medchemexpress LLC KHK-IN-1 hydrochloride | 1303470-48-5 | 98.5% | 1 ML
SDP
Catalog No. 5000339107
Encompass_Preferred
KHK-IN-1 hydrochloride (compound 8) is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor with an IC50 of 12 nM and F=34%. It inhibits the production of F1P in HepG2 cell lysates with an IC50 of 400 nM. This compound has potential for the study of diabetes and obesity.
  • Selective and cell membrane permeable KHK inhibitor
  • Inhibits F1P production in HepG2 cell lysates
  • Potential for the study of diabetes and obesity
  • Stable in human and rat liver microsome preparations
  • Does not significantly inhibit cytochrome P450s
  • Has reasonable oral bioavailability in rats
  • Soluble in DMSO and H2O
Catalog No. 50-003-39107 Supplier Medchemexpress LLC Supplier No. HY12841A10MM/1ML
KHK-IN-1 hydrochloride (compound 8) is a selective and cell membrane permeable ketohexokinase (KHK) inhibitor with an IC50 of 12 nM and F=34%. It inhibits the production of F1P in HepG2 cell lysates with an IC50 of 400 nM. This compound has potential for the study of diabetes and obesity.
  • Selective and cell membrane permeable KHK inhibitor
  • Inhibits F1P production in HepG2 cell lysates
  • Potential for the study of diabetes and obesity
  • Stable in human and rat liver microsome preparations
  • Does not significantly inhibit cytochrome P450s
  • Has reasonable oral bioavailability in rats
  • Soluble in DMSO and H2O