Learn More
Medchemexpress LLC KHK-IN-2 | 2135304-43-5 | 99.1% | 372.3 g/mol | C16H19F3N4O3 | 50 MG
SDP
Catalog No. 5000249202
Encompass_Preferred
KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor used as a research tool to probe KHK activity and related metabolic pathways. It has an IC50 of 0.45 μM against KHK and is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent and selective KHK inhibitor.
  • IC50 0.45 μM against KHK.
  • High purity (≈99.1%) suitable for research use.
  • Supplied as a solid; commonly dissolved in DMSO for assays.
  • Available in small milligram quantities for screening and mechanistic studies.
  • Applicable to enzymology, cellular assays, and metabolic research.
Catalog No. 50-002-49202 Supplier Medchemexpress LLC Supplier No. HY10347850MG
KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor used as a research tool to probe KHK activity and related metabolic pathways. It has an IC50 of 0.45 μM against KHK and is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent and selective KHK inhibitor.
  • IC50 0.45 μM against KHK.
  • High purity (≈99.1%) suitable for research use.
  • Supplied as a solid; commonly dissolved in DMSO for assays.
  • Available in small milligram quantities for screening and mechanistic studies.
  • Applicable to enzymology, cellular assays, and metabolic research.